Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Oxabicyclo[3.1.0]hexane-2,4-dione, ≥97%

Chemical Identifiers

Chemical Name or Material 3-Oxabicyclo[3.1.0]hexane-2,4-dione
CAS 5617-74-3
Formula Weight 112.08
MDL Number MFCD00126929
Beilstein 113493
Synonym 1,2-Cyclopropanedicarboxylic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6438203

  SDS
alfa aesar™
H6438203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64382MD

  SDS
alfa aesar™
H64382MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 100° to 102°C (5mmHg)
Formula Weight 112.08
Melting Point 59° to 61°C (lit.)
Synonym 1,2-Cyclopropanedicarboxylic anhydride
Chemical Name or Material 3-Oxabicyclo[3.1.0]hexane-2,4-dione
CAS 5617-74-3
Color White
Physical Form Powder
Density 1.50g/mL
Sensitivity Moisture Sensitive
Assay ≥97%
Beilstein 113493
Quantity 1g
MDL Number MFCD00126929