Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Octyne, 98%

Chemical Identifiers

Chemical Name or Material 3-Octyne
CAS 15232-76-5
Molecular Formula C8H14
Linear Formula CH3(CH2)3C≡CCH2CH3
Formula Weight 110.20g/mol
MDL Number MFCD00027276
UN Number UN3295
Synonym n-Butyl ethyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1284503

 
alfa aesar™
A12845-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 133°C
Melting Point −104°C
Quantity 1g
Flash Point 24°C (75°F)
Odor Petroleum-like
Chemical Name or Material 3-Octyne
CAS 15232-76-5
Assay 98%
Molecular Formula C8H14
Linear Formula CH3(CH2)3C≡CCH2CH3
Formula Weight 110.20g/mol
MDL Number MFCD00027276
UN Number UN3295
Synonym n-Butyl ethyl acetylene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.