missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Octylthiophene, 94%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 296140250

 View more versions of this product

Catalog No. AC296140250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Octylthiophene
65016-62-8
100.0
0.9100g/mL
Glass bottle
≥92%
1.4890 to 1.4950 (20°C, 589nm)
196.36
Clear colorless to light yellow
94%
92.0
remainder mainly tetradecane
Authentic
Liquid
97%
C12H20S
MFCD00085281
25g

Chemical Identifiers

65016-62-8
100.0
C12H20S
MFCD00085281
≥92%
92.0
97%
196.36
3-Octylthiophene, 94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA