missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Octen-2-one, 97%

Manufacturer:  Alfa Aesar™ A1465230

 View more versions of this product

Catalog No. AAA1465230



Specifications

Specifications

3-Octen-2-one
Pleasant
250g
C8H14O
UN1224
3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
CCCCC=CC(=O)C
126.199
126.2
54°C (129°F)
60°C (16mmHg)
1669-44-9
MFCD00015565
1742230
ZCFOBLITZWHNNC-VOTSOKGWSA-N
(E)-oct-3-en-2-one
5363229
97%

Chemical Identifiers

1669-44-9
126.199
ZCFOBLITZWHNNC-VOTSOKGWSA-N
5363229
CCCCC=CC(=O)C
C8H14O
MFCD00015565
3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
(E)-oct-3-en-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA

Documents