Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Octen-2-ol, 97%

$88.36 - $317.09

Chemical Identifiers

Chemical Name or Material 3-Octen-2-ol
CAS 76649-14-4
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD00036565
Beilstein 1743648
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2174106

 
alfa aesar™
B2174106
5g Each for $88.36

AAB2174114

 
alfa aesar™
B2174114
25g Each for $317.09
Description & Specifications

Specifications

Formula Weight 128.22
Chemical Name or Material 3-Octen-2-ol
TSCA TSCA
Assay 97%
Molecular Formula C8H16O
MDL Number MFCD00036565
CAS 76649-14-4
Beilstein 1743648
Boiling Point 98°C (2mmHg)
Quantity 5g
Flash Point 71°C (159°F)