missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Octadecylthiophene 98.0+%, TCI America™
$78.29 - $238.60
Chemical Identifiers
| CAS | 104934-54-5 |
|---|---|
| Molecular Formula | C22H40S |
| Molecular Weight (g/mol) | 336.622 |
| MDL Number | MFCD00191567 |
| InChI Key | ARFJPHXJBIEWSZ-UHFFFAOYSA-N |
| Synonym | 3-Stearylthiophene |
| PubChem CID | 566857 |
| IUPAC Name | 3-octadecylthiophene |
| SMILES | CCCCCCCCCCCCCCCCCCC1=CSC=C1 |
Chemical Identifiers
| 104934-54-5 | |
| 336.622 | |
| ARFJPHXJBIEWSZ-UHFFFAOYSA-N | |
| 566857 | |
| CCCCCCCCCCCCCCCCCCC1=CSC=C1 |
| C22H40S | |
| MFCD00191567 | |
| 3-Stearylthiophene | |
| 3-octadecylthiophene |
Specifications
| 104934-54-5 | |
| White | |
| MFCD00191567 | |
| 3-Stearylthiophene | |
| CCCCCCCCCCCCCCCCCCC1=CSC=C1 | |
| 336.622 | |
| 336.62 | |
| Crystalline Powder |
| 36°C | |
| C22H40S | |
| 1 g | |
| ARFJPHXJBIEWSZ-UHFFFAOYSA-N | |
| 3-octadecylthiophene | |
| 566857 | |
| ≥98.0% (GC) | |
| 3-Octadecylthiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator