missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-O-tert-Butyldimethylsilyl-4,6-O-isopropylidene-D-glucal, 97%

Manufacturer:  Alfa Aesar™ H60072MD

 View more versions of this product

Catalog No. AAH60072MD



Specifications

Specifications

3-O-tert-Butyldimethylsilyl-4,6-O-isopropylidene-D-glucal
121846-11-5
MFCD22988990
PYHWHKQRALSXOY-UPJWGTAASA-N
[(4aR,8R,8aR)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy-tert-butyl-dimethylsilane
14352117
97%
250mg
C15H28O4Si
3-o-tert-butyldimethylsilyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8ar-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
CC1(OCC2C(O1)C(C=CO2)O[Si](C)(C)C(C)(C)C)C
300.47
300.47

Chemical Identifiers

121846-11-5
300.47
PYHWHKQRALSXOY-UPJWGTAASA-N
14352117
CC1(OCC2C(O1)C(C=CO2)O[Si](C)(C)C(C)(C)C)C
C15H28O4Si
MFCD22988990
3-o-tert-butyldimethylsilyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8ar-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
[(4aR,8R,8aR)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy-tert-butyl-dimethylsilane
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents