missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-O-tert-Butyldimethylsilyl-4,6-O-(4-methoxybenzylidene)-D-glucal, 97%

$50.12 - $158.84

Chemical Identifiers

CAS 384346-91-2
Molecular Formula C20H30O5Si
Molecular Weight (g/mol) 378.54
MDL Number MFCD01863523
InChI Key ZTRSSQRWGPRALW-KAKFPZCNSA-N
Synonym 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
PubChem CID 71311532
IUPAC Name [(4aR,8R,8aR)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1C=COC2C1OC(OC2)C3=CC=C(C=C3)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60924MD View Documents Alfa Aesar™
H60924MD
250mg Each for $50.12
 
AAH6092403 View Documents Alfa Aesar™
H6092403
1g Each for $158.84
 
Specifications

Chemical Identifiers

384346-91-2
378.54
ZTRSSQRWGPRALW-KAKFPZCNSA-N
71311532
CC(C)(C)[Si](C)(C)OC1C=COC2C1OC(OC2)C3=CC=C(C=C3)OC
C20H30O5Si
MFCD01863523
3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
[(4aR,8R,8aR)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy-tert-butyl-dimethylsilane

Specifications

250mg
C20H30O5Si
3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
CC(C)(C)[Si](C)(C)OC1C=COC2C1OC(OC2)C3=CC=C(C=C3)OC
378.54
378.53
3-O-tert-Butyldimethylsilyl-4,6-O-(4-methoxybenzylidene)-D-glucal
384346-91-2
MFCD01863523
ZTRSSQRWGPRALW-KAKFPZCNSA-N
[(4aR,8R,8aR)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy-tert-butyl-dimethylsilane
71311532
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C