Please login to your online account to display your discounted pricing

3-O-Methyldopamine hydrochloride, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  172850010

 View more versions of this product

Catalog No. AC172850010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥99%
Formula Weight 203.67
Quantity 1g
Color Slightly beige
EINECS Number 216-035-1
TSCA TSCA
CAS Max % 100.0
CAS Min % 99.0
Chemical Name or Material 3-O-Methyldopamine hydrochloride
Name Note p.a.
Assay Percent Range 99+%
Molecular Formula C9H13NO2·HCl
MDL Number MFCD00012902
CAS 1477-68-5
Synonym 3-Methoxytyramine hydrochloride
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-O-Methyldopamine hydrochloride
Name Note p.a.
CAS 1477-68-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99%
Assay Percent Range 99+%
Molecular Formula C9H13NO2·HCl
Formula Weight 203.67
MDL Number MFCD00012902
Synonym 3-Methoxytyramine hydrochloride
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents