missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-O-Ethyl-L-ascorbic Acid 98.0+%, TCI America™
Supplier: TCI America E092625G
Specifications
3-O-Ethyl-L-ascorbic Acid | |
114°C | |
C8H12O6 | |
25 g | |
CCOC1=C(C(=O)OC1C(CO)O)O | |
204.178 | |
204.18 | |
Crystalline Powder |
86404-04-8 | |
White | |
MFCD09261382 | |
ZGSCRDSBTNQPMS-UJURSFKZSA-N | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one | |
150736 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
86404-04-8 | |
204.178 | |
ZGSCRDSBTNQPMS-UJURSFKZSA-N | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one |
C8H12O6 | |
MFCD09261382 | |
150736 | |
CCOC1=C(C(=O)OC1C(CO)O)O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator