missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-O-Benzyl-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose, 98%

$33.01 - $215.89

Chemical Identifiers

CAS 18685-18-2
Molecular Formula C19H26O6
Molecular Weight (g/mol) 350.411
MDL Number MFCD00269929
InChI Key ZHFVGOMEUGAIJX-QTPCEWDRSA-N
Synonym 3-o-benzyl-1,2:5,6-di-o-isopropylidene-, a-d-glucofuranose, 1,2:5,6-bis-o-1-methylethylidene-3-o-phenylmethyl-, a-d-glucofuranose, 4-3ar,5r,6as-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl-2,2-dimethyl-1,3-dioxolane
PubChem CID 45038272
IUPAC Name (3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)OCC4=CC=CC=C4)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66578ME View Documents Alfa Aesar™
J66578ME
500mg Each for $33.01
 
AAJ6657803 View Documents Alfa Aesar™
J6657803
1g Each for $52.43
 
AAJ6657806 View Documents Alfa Aesar™
J6657806
5g Each for $215.89
 
Specifications

Chemical Identifiers

18685-18-2
350.411
ZHFVGOMEUGAIJX-QTPCEWDRSA-N
45038272
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)OCC4=CC=CC=C4)C
C19H26O6
MFCD00269929
3-o-benzyl-1,2:5,6-di-o-isopropylidene-, a-d-glucofuranose, 1,2:5,6-bis-o-1-methylethylidene-3-o-phenylmethyl-, a-d-glucofuranose, 4-3ar,5r,6as-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl-2,2-dimethyl-1,3-dioxolane
(3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole

Specifications

1.094
1.4925
Colorless to Yellow
18685-18-2
MFCD00269929
ZHFVGOMEUGAIJX-QTPCEWDRSA-N
(3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-6-phenylmethoxy-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole
45038272
Oil
3-O-Benzyl-1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose
−30° (c=1 in Ethanol)
140°C to 142°C
500mg
C19H26O6
3-o-benzyl-1,2:5,6-di-o-isopropylidene-, a-d-glucofuranose, 1,2:5,6-bis-o-1-methylethylidene-3-o-phenylmethyl-, a-d-glucofuranose, 4-3ar,5r,6as-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl-2,2-dimethyl-1,3-dioxolane
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)OCC4=CC=CC=C4)C
350.411
350.41
98%
Safety and Handling

Safety and Handling

RTECSNumber : LZ4918000