missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-O-Benzoyl-4,6-O-isopropylidene-D-glucal, 97%

$55.51 - $175.80

Chemical Identifiers

CAS 58871-20-8
Molecular Formula C16H18O5
Molecular Weight (g/mol) 290.315
MDL Number MFCD22988992
InChI Key RADJUQJHMUFUJI-MCIONIFRSA-N
Synonym 3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
PubChem CID 92135621
IUPAC Name [(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate
SMILES CC1(OCC2C(O1)C(C=CO2)OC(=O)C3=CC=CC=C3)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60142MD View Documents Alfa Aesar™
H60142MD
250mg Each for $55.51
 
AAH6014203 View Documents Alfa Aesar™
H6014203
1g Each for $175.80
 
Specifications

Chemical Identifiers

58871-20-8
290.315
RADJUQJHMUFUJI-MCIONIFRSA-N
92135621
CC1(OCC2C(O1)C(C=CO2)OC(=O)C3=CC=CC=C3)C
C16H18O5
MFCD22988992
3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
[(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate

Specifications

250mg
C16H18O5
3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
CC1(OCC2C(O1)C(C=CO2)OC(=O)C3=CC=CC=C3)C
290.315
290.31
3-O-Benzoyl-4,6-O-isopropylidene-D-glucal
58871-20-8
MFCD22988992
RADJUQJHMUFUJI-MCIONIFRSA-N
[(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate
92135621
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C