missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-O-Acetyl-4,6-O-isopropylidene-D-glucal, 97%

$56.74 - $178.89

Chemical Identifiers

CAS 97747-17-6
Molecular Formula C11H16O5
Molecular Weight (g/mol) 228.244
MDL Number MFCD22989002
InChI Key JSKNNUBJVJGXIN-BBBLOLIVSA-N
Synonym 3-o-acetyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl acetate, 1,5-anhydro-2-deoxy-3-o-acetyl-4-o,6-o-isopropylidene-d-arabino-hexa-1-enitol
PubChem CID 25259938
IUPAC Name [(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] acetate
SMILES CC(=O)OC1C=COC2C1OC(OC2)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60386MD View Documents Alfa Aesar™
H60386MD
250mg Each for $56.74
 
AAH6038603 View Documents Alfa Aesar™
H6038603
1g Each for $178.89
 
Specifications

Chemical Identifiers

97747-17-6
228.244
JSKNNUBJVJGXIN-BBBLOLIVSA-N
25259938
CC(=O)OC1C=COC2C1OC(OC2)(C)C
C11H16O5
MFCD22989002
3-o-acetyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl acetate, 1,5-anhydro-2-deoxy-3-o-acetyl-4-o,6-o-isopropylidene-d-arabino-hexa-1-enitol
[(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] acetate

Specifications

250mg
C11H16O5
3-o-acetyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl acetate, 1,5-anhydro-2-deoxy-3-o-acetyl-4-o,6-o-isopropylidene-d-arabino-hexa-1-enitol
CC(=O)OC1C=COC2C1OC(OC2)(C)C
228.244
228.24
3-O-Acetyl-4,6-O-isopropylidene-D-glucal
97747-17-6
MFCD22989002
JSKNNUBJVJGXIN-BBBLOLIVSA-N
[(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] acetate
25259938
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C