Please login to your online account to display your discounted pricing

3-Noradamantanecarboxylic acid, 98%, ACROS Organics™

$147.54 - $147.54

Chemical Identifiers

Chemical Name or Material 3-Noradamantanecarboxylic acid
Name Note 98%
CAS 16200-53-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H14O2
Formula Weight 166.22
MDL Number MFCD00213463
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC339280010

  SDS
thermo fisher scientific
339280010
Glass bottle 1g Each for $147.54
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 166.22
Physical Form Crystalline Powder and Chunks or Crystals
Quantity 1g
Color White
TSCA TSCA
Melting Point 104° to 108°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Chemical Name or Material 3-Noradamantanecarboxylic acid
Assay Percent Range 98%
CAS 16200-53-6
Name Note 98%
Molecular Formula C10H14O2
MDL Number MFCD00213463
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.