missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Noradamantanamine Hydrochloride, TCI America™

$262.78 - $873.91

Chemical Identifiers

CAS 86128-83-8
Molecular Formula C9H16ClN
Molecular Weight (g/mol) 173.684
MDL Number MFCD00213499
InChI Key YPEGOYFGBDTDJK-UHFFFAOYSA-N
Synonym 3-noradamantanamine hydrochloride, octahydro-2,5-methanopentalen-3a-amine hydrochloride, 3-aminonoradamantane hcl, 3-aminonoradamantane hydrochloride, tricyclo 3.3.1.0 3 ,? nonan-3-amine hydrochloride, 3-noradamantanamine hcl, c9h15n.hcl, 3-noradamantanaminehydrochloride, 3-noradamantanamine hydrochloride 95, 3-noradamantanamine hydrochloride
PubChem CID 12791864
SMILES C1C2CC3CC1CC3(C2)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N07781G View Documents TCI America
N07781G
1g Each for $262.78
 
N07785G View Documents TCI America
N07785G
5g Each for $873.91
 
Specifications

Chemical Identifiers

86128-83-8
173.684
YPEGOYFGBDTDJK-UHFFFAOYSA-N
12791864
C9H16ClN
MFCD00213499
3-noradamantanamine hydrochloride, octahydro-2,5-methanopentalen-3a-amine hydrochloride, 3-aminonoradamantane hcl, 3-aminonoradamantane hydrochloride, tricyclo 3.3.1.0 3 ,? nonan-3-amine hydrochloride, 3-noradamantanamine hcl, c9h15n.hcl, 3-noradamantanaminehydrochloride, 3-noradamantanamine hydrochloride 95, 3-noradamantanamine hydrochloride
C1C2CC3CC1CC3(C2)N.Cl

Specifications

White-Yellow
1g
C9H16ClN
3-noradamantanamine hydrochloride, octahydro-2,5-methanopentalen-3a-amine hydrochloride, 3-aminonoradamantane hcl, 3-aminonoradamantane hydrochloride, tricyclo 3.3.1.0 3 ,? nonan-3-amine hydrochloride, 3-noradamantanamine hcl, c9h15n.hcl, 3-noradamantanaminehydrochloride, 3-noradamantanamine hydrochloride 95, 3-noradamantanamine hydrochloride
C1C2CC3CC1CC3(C2)N.Cl
12791864
Crystal-Powder at 20°C
3-Noradamantanamine Hydrochloride
350°C
86128-83-8
MFCD00213499
YPEGOYFGBDTDJK-UHFFFAOYSA-N
173.684
173.68
≥98.0% (N,T)
Safety and Handling

Safety and Handling

TSCA : No