Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nonyn-1-ol, 98%

$76.85 - $76.85

Chemical Identifiers

Chemical Name or Material 3-Nonyn-1-ol
CAS 31333-13-8
Molecular Formula CH3(CH2)4C≡CCH2CH2OH
Formula Weight 140.23
MDL Number MFCD00002958
Beilstein 1700444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1292206

  SDS
alfa aesar™
A1292206
5g Each for $76.85

AAA1292209

  SDS
alfa aesar™
A1292209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-Nonyn-1-ol
Flash Point 93°C (199°F)
Boiling Point 111° to 112°C (20mmHg)
Quantity 5g
CAS 31333-13-8
Assay 98%
Molecular Formula CH3(CH2)4C≡CCH2CH2OH
Formula Weight 140.23
MDL Number MFCD00002958
Beilstein 1700444

Applications
It is used in the synthesis of 3,3,4,4- 2 H 4 -nonanol and 3,3,4,4- 2 H 4 -nonanal.

Solubility
Optimally soluble in aqueous solutions.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.