Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nonanone, 98%

$56.50 - $123.41

Chemical Identifiers

Chemical Name or Material 3-Nonanone
CAS 925-78-0
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00009541
Beilstein 1700447
Synonym Ethyl n-hexyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1987814

  SDS
alfa aesar™
A1987814
25g Each for $56.50

AAA1987822

  SDS
alfa aesar™
A1987822
100g Each for $123.41
Description & Specifications

Specifications

Formula Weight 142.24
Chemical Name or Material 3-Nonanone
TSCA TSCA
Assay 98%
Molecular Formula C9H18O
MDL Number MFCD00009541
CAS 925-78-0
Beilstein 1700447
Synonym Ethyl n-hexyl ketone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 63°C (145°F)
Boiling Point 187° to 188°C
Melting Point −8°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.