Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitropyridine, 97%

$170.27 - $170.27

Chemical Identifiers

Chemical Name or Material 3-Nitropyridine
CAS 2530-26-9
Molecular Formula C5H4N2O2
Formula Weight 124.1
MDL Number MFCD00234974
UN Number UN2811
Beilstein 111969
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1962706

  SDS
alfa aesar™
L1962706
5g Each for $170.27

AAL1962703

  SDS
alfa aesar™
L1962703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1962714

  SDS
alfa aesar™
L1962714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.1
Chemical Name or Material 3-Nitropyridine
Melting Point 38° to 42°C
Boiling Point 216°C
Assay 97%
Quantity 5g
Molecular Formula C5H4N2O2
MDL Number MFCD00234974
CAS 2530-26-9
UN Number UN2811
Beilstein 111969
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.