Alfa Aesar™ 3'-Nitropropiophenone, 97%
Manufacturer:  Alfa Aesar™ A1455422
 View more versions of this product
Catalog No. AAA1455422
Specifications
Specifications
3'-Nitropropiophenone
100g
C9H9NO3
1954069
VSPOTMOYDHRALZ-UHFFFAOYSA-N
1-(3-nitrophenyl)propan-1-one
87096
97%
99°C to 102°C
17408-16-1
MFCD00051516
3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one
CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
179.175
179.18
Chemical Identifiers
17408-16-1
179.175
VSPOTMOYDHRALZ-UHFFFAOYSA-N
87096
CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
C9H9NO3
MFCD00051516
3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one
1-(3-nitrophenyl)propan-1-one
Documents