Please login to your online account to display your discounted pricing

3-Nitrophthalonitrile, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Nitrophthalonitrile
Name Note 99%
CAS 51762-67-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8 H3 N3 O2
Formula Weight 173.13
MDL Number MFCD00191558
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC360250010

  SDS
thermo fisher scientific
AC360250010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 173.13
Physical Form Powder
Quantity 1g
Color Light yellow
Melting Point 162° to 165°C
CAS Max % 100.0
Name Note 99%
Chemical Name or Material 3-Nitrophthalonitrile
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 51762-67-5
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00191558
Molecular Formula C8 H3 N3 O2
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.