Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrophthalonitrile, 99%

$94.93 - $278.76

Chemical Identifiers

Chemical Name or Material 3-Nitrophthalonitrile
CAS 51762-67-5
Molecular Formula C8H3N3O2
Formula Weight 173.13
MDL Number MFCD00191558
Beilstein 2263686
Synonym 2,3-Dicyanonitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2526906

  SDS
alfa aesar™
B2526906
5g Each for $94.93

AAB2526914

  SDS
alfa aesar™
B2526914
25g Each for $278.76
Description & Specifications

Specifications

Formula Weight 173.13
Assay 99%
Synonym 2,3-Dicyanonitrobenzene
Health Hazard 1 Warning
CAS 51762-67-5
RTECS Number CZ1953200
MDL Number MFCD00191558
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 162° to 165°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Chemical Name or Material 3-Nitrophthalonitrile
Beilstein 2263686
Molecular Formula C8H3N3O2

Applications
3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incombatible with strong oxidizing agents and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.