Learn More
3-Nitrophthalimide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1529122
Description
3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Nitrophthalimide is a useful dye for biological research purposes. It is a nitro heterocyclic compounds found to exhibit potential antifungal activities.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents and bases.
Specifications
3-Nitrophthalimide | |
217°C to 221°C | |
MFCD00041852 | |
179965 | |
Insoluble in water. | |
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=O)NC2=O | |
192.13 | |
192.13 |
603-62-3 | |
C8H4N2O4 | |
100 g | |
3-nitrophthalimide, 4-nitroisoindoline-1,3-dione, phthalimide, 3-nitro, 1h-isoindole-1,3 2h-dione, 4-nitro, nitrophthalimide, 4-nitro-2,3-dihydro-1h-isoindole-1,3-dione, 4-nitro-1h-isoindole-1,3 2h-dione, 4-nitro-2h-benzo c azoline-1,3-dione, 3-nitro-phthalimide, pubchem12007 | |
BONIIQYTWOPUQI-UHFFFAOYSA-N | |
4-nitroisoindole-1,3-dione | |
11779 | |
98% |
Chemical Identifiers
603-62-3 | |
192.13 | |
BONIIQYTWOPUQI-UHFFFAOYSA-N | |
11779 | |
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=O)NC2=O |
C8H4N2O4 | |
MFCD00041852 | |
3-nitrophthalimide, 4-nitroisoindoline-1,3-dione, phthalimide, 3-nitro, 1h-isoindole-1,3 2h-dione, 4-nitro, nitrophthalimide, 4-nitro-2,3-dihydro-1h-isoindole-1,3-dione, 4-nitro-1h-isoindole-1,3 2h-dione, 4-nitro-2h-benzo c azoline-1,3-dione, 3-nitro-phthalimide, pubchem12007 | |
4-nitroisoindole-1,3-dione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-051-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only