Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrophenylsulfur pentafluoride, 95%

Chemical Identifiers

Chemical Name or Material 3-Nitrophenylsulfur pentafluoride
CAS 2613-26-5
Molecular Formula C6H4F5NO2S
Formula Weight 249.16
MDL Number MFCD00221621
UN Number UN2810
Beilstein 2699206
Synonym 3-Nitrobenzenesulfur pentafluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1754503

  SDS
alfa aesar™
L1754503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.16
Assay 95%
Molecular Formula C6H4F5NO2S
MDL Number MFCD00221621
CAS 2613-26-5
Beilstein 2699206
Synonym 3-Nitrobenzenesulfur pentafluoride
Chemical Name or Material 3-Nitrophenylsulfur pentafluoride
UN Number UN2810
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Boiling Point 106°C (2mmHg)
Quantity 1g

Applications
It has a wide range of applications as a fluorinating agent. g. It is a convenient alternative to current fluorinating agents for use in the synthesis of specialty chemicals and intermediates, such as pharmaceuticals, agrochemicals and electronics chemicals.

Solubility
Difficult to mix in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.