missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophenylhydrazine Hydrochloride 98%, Acros Organics

Chemical Identifiers

CAS 636-95-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H7N3O2·HCl
Linear Formula O2NC6H4NHNH2·HCl
Formula Weight 189.6
MDL Number MFCD00012939
Chemical Name or Material 3-Nitrophenylhydrazine hydrochloride, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC128840250 SDS Thermo Fisher Scientific
128840250
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

636-95-3
100.0
C6H7N3O2·HCl
189.6
3-Nitrophenylhydrazine hydrochloride, 98%
97.5
98%
O2NC6H4NHNH2·HCl
MFCD00012939
≥97.5%

Specifications

189.6
25g
206° to 210°C
100.0
Authentic
636-95-3
C6H7N3O2·HCl
O2NC6H4NHNH2·HCl
≥97.5%
Crystalline Powder
Yellow
15, 460
0.05% max.
97.5
98%
MFCD00012939
3-Nitrophenylhydrazine hydrochloride, 98%