Please login to your online account to display your discounted pricing

3-Nitrophenylhydrazine Hydrochloride 98%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  128840250

 View more versions of this product

Catalog No. AC128840250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 189.6
Physical Form Crystalline Powder
Quantity 25g
Color Yellow
TSCA TSCA
EINECS Number 211-270-6
Melting Point 206° to 210°C
Beilstein 15, 460
CAS Max % 100.0
Sulfated Ash 0.05% max.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Risk of explosion if heated under confinement.
CAS 636-95-3
Chemical Name or Material 3-Nitrophenylhydrazine hydrochloride
Assay Percent Range 98%
Molecular Formula C6H7N3O2·HCl
MDL Number MFCD00012939
Name Note 98%
Linear Formula O2NC6H4NHNH2·HCl

Specifications

Chemical Name or Material 3-Nitrophenylhydrazine hydrochloride
Name Note 98%
CAS 636-95-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7N3O2·HCl
Linear Formula O2NC6H4NHNH2·HCl
Formula Weight 189.6
MDL Number MFCD00012939
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Risk of explosion if heated under confinement.

SDS & Safety Documents