missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophenylacetic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC147050250

 View more versions of this product

Catalog No. AC147050250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Nitrophenylacetic acid
Glass bottle
116.0°C to 120.0°C
1877-73-2
100.0
C8H7NO4
MFCD00007278
2-3-nitrophenyl acetic acid, 3-nitrobenzeneacetic acid, 3-nitrophenyl acetic acid, 3-nitrophenylacetic acid, acetic acid, m-nitrophenyl, benzeneacetic acid, 3-nitro, ccris 2337, m-nitrophenyl acetic acid, m-nitrophenylacetic acid
C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O
15876
Crystalline Powder
Authentic
Cream
25g
98.5
98.5% min. (GC)
O2NC6H4CH2CO2H
09, 455
WUKHOVCMWXMOOA-UHFFFAOYSA-N
181.15
181.15
99%

Chemical Identifiers

1877-73-2
181.15
WUKHOVCMWXMOOA-UHFFFAOYSA-N
15876
C8H7NO4
MFCD00007278
2-3-nitrophenyl acetic acid, 3-nitrobenzeneacetic acid, 3-nitrophenyl acetic acid, 3-nitrophenylacetic acid, acetic acid, m-nitrophenyl, benzeneacetic acid, 3-nitro, ccris 2337, m-nitrophenyl acetic acid, m-nitrophenylacetic acid
C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-512-7

SDS Documents