missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophenylacetic acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics 147050050

 View more versions of this product

Catalog No. AC147050050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Nitrophenylacetic acid
116°C to 120°C
1877-73-2
100.0
Glass bottle
99%
O2NC6H4CH2CO2H
09, 455
C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O
15876
Crystalline Powder
Cream
5g
98.5
Authentic
≥98.5%
C8H7NO4
MFCD00007278
WUKHOVCMWXMOOA-UHFFFAOYSA-N
181.147
181.15

Chemical Identifiers

1877-73-2
181.147
WUKHOVCMWXMOOA-UHFFFAOYSA-N
C8H7NO4
MFCD00007278
15876
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents