missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrophenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1133503

 View more versions of this product

Catalog No. AAL1133503



Specifications

Specifications

3-Nitrophenyl isothiocyanate
56°C to 58°C
3529-82-6
MFCD00041245
2094061
OEZXLKSZOAWNJU-UHFFFAOYSA-N
1-isothiocyanato-3-nitrobenzene
19050
97%
Irritating
1g
C7H4N2O2S
UN2923
3-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-3-nitro, m-nitrophenyl isothiocyanate, 3-nitrophenylisothiocyanate, isothiocyanic acid, m-nitrophenyl ester, 1-isothiocyanato-3-nitro-benzene, m-nitrophenylisothiocyanat, acmc-1co3y, phenyl, 3-nitro, isothiocyanate, #
C1=CC(=CC(=C1)[N+](=O)[O-])N=C=S
180.181
180.19

Chemical Identifiers

3529-82-6
180.181
OEZXLKSZOAWNJU-UHFFFAOYSA-N
19050
C1=CC(=CC(=C1)[N+](=O)[O-])N=C=S
C7H4N2O2S
MFCD00041245
3-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-3-nitro, m-nitrophenyl isothiocyanate, 3-nitrophenylisothiocyanate, isothiocyanic acid, m-nitrophenyl ester, 1-isothiocyanato-3-nitro-benzene, m-nitrophenylisothiocyanat, acmc-1co3y, phenyl, 3-nitro, isothiocyanate, #
1-isothiocyanato-3-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NX9110000

TSCA : TSCA

Documents