missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrophenoxyacetic Acid, TCI America™

$168.06 - $455.28

Chemical Identifiers

CAS 1878-88-2
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00059826
InChI Key BNRRQAASFDGMMQ-UHFFFAOYSA-N
Synonym 3-nitrophenoxyacetic acid, 3-nitrophenoxy acetic acid, 2-3-nitrophenoxy acetic acid, m-nitrophenoxyacetic acid, 3-nitrophenoxy-acetic acid, acmc-209eqk, 3-nitrophenoxyaceticacid, 3-nitrophenoxy acetic acid, 3-nitrophenoxy acetic acid, 3-nitro-phenoxy-acetic acid
PubChem CID 74656
SMILES C1=CC(=CC(=C1)OCC(=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N022425G View Documents TCI America
N022425G
25g Each for $455.28
 
N02245G View Documents TCI America
N02245G
5g Each for $168.06
 
Specifications

Chemical Identifiers

1878-88-2
197.146
BNRRQAASFDGMMQ-UHFFFAOYSA-N
74656
C8H7NO5
MFCD00059826
3-nitrophenoxyacetic acid, 3-nitrophenoxy acetic acid, 2-3-nitrophenoxy acetic acid, m-nitrophenoxyacetic acid, 3-nitrophenoxy-acetic acid, acmc-209eqk, 3-nitrophenoxyaceticacid, 3-nitrophenoxy acetic acid, 3-nitrophenoxy acetic acid, 3-nitro-phenoxy-acetic acid
C1=CC(=CC(=C1)OCC(=O)O)[N+](=O)[O-]

Specifications

Yellow
25g
C8H7NO5
3-nitrophenoxyacetic acid, 3-nitrophenoxy acetic acid, 2-3-nitrophenoxy acetic acid, m-nitrophenoxyacetic acid, 3-nitrophenoxy-acetic acid, acmc-209eqk, 3-nitrophenoxyaceticacid, 3-nitrophenoxy acetic acid, 3-nitrophenoxy acetic acid, 3-nitro-phenoxy-acetic acid
C1=CC(=CC(=C1)OCC(=O)O)[N+](=O)[O-]
74656
Crystal-Powder at 20°C
3-Nitrophenoxyacetic Acid
153°C
1878-88-2
MFCD00059826
BNRRQAASFDGMMQ-UHFFFAOYSA-N
197.146
197.15
≥98.0% (T,GC)
Safety and Handling

Safety and Handling

TSCA : No