missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrophenol, 98+%

$94.51 - $995.92

Chemical Identifiers

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym 1-hydroxy-3-nitrobenzene, 3-hydroxynitrobenzene, 3-nitro-phenol, m-hydroxynitrobenzene, m-nitrofenol, m-nitrofenol czech, m-nitrophenol, meta-nitrophenol, phenol, 3-nitro, phenol, m-nitro
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES C1=CC(=CC(=C1)O)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1531209
View Documents
Alfa Aesar™
A1531209
10g
Each for $94.51
 
AAA1531218
View Documents
Alfa Aesar™
A1531218
50g
Each for $243.03
 
AAA1531230
View Documents
Alfa Aesar™
A1531230
250g
Each for $995.92
 
Description

Description

Applications
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).

Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.
Specifications

Chemical Identifiers

554-84-7
139.11
RTZZCYNQPHTPPL-UHFFFAOYSA-N
11137
3-nitrophenol
C6H5NO3
MFCD00007240
1-hydroxy-3-nitrobenzene, 3-hydroxynitrobenzene, 3-nitro-phenol, m-hydroxynitrobenzene, m-nitrofenol, m-nitrofenol czech, m-nitrophenol, meta-nitrophenol, phenol, 3-nitro, phenol, m-nitro
CHEBI:34346
C1=CC(=CC(=C1)O)[N+](=O)[O-]

Specifications

1.49
194°C (70mmHg)
10g
C6H5NO3
UN1663
14,6619
Soluble in water (30mg/ml),ether,alcohol,acetone,and ethanol (700mg/ml).
C1=CC(=CC(=C1)O)[N+](=O)[O-]
139.11
CHEBI:34346
≥98%
>100°C (212°F)
93°C to 98°C
554-84-7
MFCD00007240
1907946
1-hydroxy-3-nitrobenzene, 3-hydroxynitrobenzene, 3-nitro-phenol, m-hydroxynitrobenzene, m-nitrofenol, m-nitrofenol czech, m-nitrophenol, meta-nitrophenol, phenol, 3-nitro, phenol, m-nitro
RTZZCYNQPHTPPL-UHFFFAOYSA-N
3-nitrophenol
11137
139.11
3-Nitrophenol
Safety and Handling

Safety and Handling

GHS H Statement
H318-H373-H302-H312-H335-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P332+P313-P362-P501c

H302-H315-H318-H373-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS

EINECSNumber : 209-073-5

RTECSNumber : SM1925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS