Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitroformanilide, 98%

$53.73 - $652.65

Chemical Identifiers

Chemical Name or Material 3-Nitroformanilide
CAS 102-38-5
Molecular Formula C7H6N2O3
Formula Weight 166.14
MDL Number MFCD00017014
Beilstein 2210047
Synonym N-(3-Nitrophenyl)formamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1527006

 
alfa aesar™
A1527006
5g Each for $53.73

AAA1527022

 
alfa aesar™
A1527022
100g Each for $652.65

AAA1527014

 
alfa aesar™
A1527014
25g Each for $197.03
Description & Specifications

Specifications

Formula Weight 166.14
Chemical Name or Material 3-Nitroformanilide
Assay 98%
Molecular Formula C7H6N2O3
MDL Number MFCD00017014
CAS 102-38-5
Beilstein 2210047
Synonym N-(3-Nitrophenyl)formamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 132° to 136°C
Quantity 5g

Applications
3-Nitroformanilide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.