missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrodiphenyl, TCI America™

$39.72 - $288.75

Chemical Identifiers

CAS 2113-58-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD00007245
InChI Key FYRPEHRWMVMHQM-UHFFFAOYSA-N
Synonym 3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
PubChem CID 16450
IUPAC Name 1-nitro-3-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N019325G View Documents TCI America
N019325G
25g Each for $288.75
 
N01931G View Documents TCI America
N01931G
1g Each for $39.72
 
Specifications

Chemical Identifiers

2113-58-8
199.209
FYRPEHRWMVMHQM-UHFFFAOYSA-N
16450
C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]
C12H9NO2
MFCD00007245
3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
1-nitro-3-phenylbenzene

Specifications

204°C
61°C
2113-58-8
MFCD00007245
FYRPEHRWMVMHQM-UHFFFAOYSA-N
1-nitro-3-phenylbenzene
16450
Crystal-Powder at 20°C
3-Nitrodiphenyl
Yellow
25g
C12H9NO2
3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]
199.209
199.21
≥99.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : DV5570000

TSCA : No