missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrocinnamic Acid 98.0+%, TCI America™

Supplier:  TCI America N035425G

 View more versions of this product

Catalog No. N035425G



Description

Description

Specifications

Specifications

3-Nitrocinnamic Acid
205°C
C9H7NO4
25 g
WWXMVRYHLZMQIG-SNAWJCMRSA-N
193.158
CHEBI:60706
≥98.0% (T)
555-68-0
White-Yellow
MFCD00007277
3-nitrocinnamic acid, m-nitrocinnamic acid, 3-nitro-cinnamic acid, cinnamic acid, m-nitro, trans-3-nitrocinnamic acid, 3-nitrobenzenepropenoic acid, e-3-3-nitrophenyl-2-propenoic acid, 2e-3-3-nitrophenyl prop-2-enoic acid, 2e-3-3-nitrophenyl acrylic acid, unii-z5wu65j930
C1=CC(=CC(=C1)[N+](=O)[O-])C=CC(=O)O
687126
193.16
Crystalline Powder

Chemical Identifiers

555-68-0
193.158
WWXMVRYHLZMQIG-SNAWJCMRSA-N
687126
C1=CC(=CC(=C1)[N+](=O)[O-])C=CC(=O)O
C9H7NO4
MFCD00007277
3-nitrocinnamic acid, m-nitrocinnamic acid, 3-nitro-cinnamic acid, cinnamic acid, m-nitro, trans-3-nitrocinnamic acid, 3-nitrobenzenepropenoic acid, e-3-3-nitrophenyl-2-propenoic acid, 2e-3-3-nitrophenyl prop-2-enoic acid, 2e-3-3-nitrophenyl acrylic acid, unii-z5wu65j930
CHEBI:60706
Safety and Handling

Safety and Handling

RTECSNumber : GE0352000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions