Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrochalcone, 98+%

$53.49 - $203.42

Chemical Identifiers

Chemical Name or Material 3-Nitrochalcone
CAS 614-48-2
Molecular Formula C15H11NO3
Formula Weight 253.26
MDL Number MFCD00024571
Beilstein 1464265
Synonym 2-(3-Nitrobenzylidene)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0115314

  SDS
alfa aesar™
L0115314
25g Each for $203.42

AAL0115306

  SDS
alfa aesar™
L0115306
5g Each for $53.49
Description & Specifications

Specifications

Formula Weight 253.26
MDL Number MFCD00024571
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 1464265
Chemical Name or Material 3-Nitrochalcone
Assay ≥98%
CAS 614-48-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Health Hazard 1 Warning
Molecular Formula C15H11NO3
Melting Point 145° to 146°C
RTECS Number FL7075490
Synonym 2-(3-Nitrobenzylidene)acetophenone
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.