missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrobiphenyl, 98+%

$53.82 - $189.68

Chemical Identifiers

CAS 2113-58-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD00007245
InChI Key FYRPEHRWMVMHQM-UHFFFAOYSA-N
Synonym 3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
PubChem CID 16450
IUPAC Name 1-nitro-3-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0124604 View Documents Alfa Aesar™
L0124604
2g Each for $53.82
 
AAL0124609 View Documents Alfa Aesar™
L0124609
10g Each for $189.68
 
Specifications

Chemical Identifiers

2113-58-8
199.209
FYRPEHRWMVMHQM-UHFFFAOYSA-N
16450
C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]
C12H9NO2
MFCD00007245
3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
1-nitro-3-phenylbenzene

Specifications

227°C (35mmHg)
2g
C12H9NO2
1872195
FYRPEHRWMVMHQM-UHFFFAOYSA-N
1-nitro-3-phenylbenzene
16450
≥98%
58°C to 60°C
2113-58-8
MFCD00007245
3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]
199.209
199.21
3-Nitrobiphenyl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DV5570000