Please login to your online account to display your discounted pricing

3-Nitrobiphenyl 97%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128570025

 View more versions of this product

Catalog No. AC128570025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 56° to 60°C
Quantity 2.5g
Color Yellow
Chemical Name or Material 3-Nitrobiphenyl
Name Note 97%
CAS 2113-58-8
CAS Min % 96.0
CAS Max % 100.0
Physical Form Yellow crystalline powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C12H9NO2
Linear Formula C6H5C6H4NO2
Formula Weight 199.21g/mol
MDL Number MFCD00007245
Beilstein 05, 582
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number DV5570000
EINECS Number 218-305-4
TSCA TSCA

Specifications

Chemical Name or Material 3-Nitrobiphenyl
Name Note 97%
CAS 2113-58-8
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C12H9NO2
Linear Formula C6H5C6H4NO2
Formula Weight 199.21g/mol
MDL Number MFCD00007245
Accessories

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.

SDS & Safety Documents