missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzyl chloride 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 404901000

 View more versions of this product

Catalog No. AC404901000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
171.58
Light yellow to light brown
100.0
Glass bottle
Authentic
619-23-8
99%
MFCD00007272
Crystalline Solid
100g
43° to 46°C
α-Chloro-m-nitrotoluene
98.5
O2NC6H4CH2Cl
3-Nitrobenzyl chloride
99%
C7H6ClNO2

Chemical Identifiers

619-23-8
100.0
C7H6ClNO2
171.58
α-Chloro-m-nitrotoluene
≥98.5%
98.5
99%
O2NC6H4CH2Cl
MFCD00007272
3-Nitrobenzyl chloride, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 210-586-1

RTECSNumber : XS9090000

TSCA : TSCA