Please login to your online account to display your discounted pricing

3-Nitrobenzyl chloride 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  404900250

 View more versions of this product

Catalog No. AC404900250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Solid
Formula Weight 171.58
Quantity 25g
Color Light yellow to light brown
TSCA TSCA
RTECS Number XS9090000
Melting Point 43° to 46°C
EINECS Number 210-586-1
CAS Max % 100.0
Synonym α-Chloro-m-nitrotoluene
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
CAS Min % 98.5
Chemical Name or Material 3-Nitrobenzyl chloride
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Linear Formula O2NC6H4CH2Cl
CAS 619-23-8
MDL Number MFCD00007272
Assay Percent Range 99%
Molecular Formula C7H6ClNO2
Name Note 99%

Specifications

Chemical Name or Material 3-Nitrobenzyl chloride
Name Note 99%
CAS 619-23-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6ClNO2
Linear Formula O2NC6H4CH2Cl
Formula Weight 171.58
MDL Number MFCD00007272
Synonym α-Chloro-m-nitrotoluene
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents