Please login to your online account to display your discounted pricing

3-Nitrobenzyl chloride 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Nitrobenzyl chloride
Name Note 99%
CAS 619-23-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6ClNO2
Linear Formula O2NC6H4CH2Cl
Formula Weight 171.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC404900250

  SDS
thermo fisher scientific
404900250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC404901000

  SDS
thermo fisher scientific
404901000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Solid
Formula Weight 171.58
Quantity 25g
Color Light yellow to light brown
EINECS Number 210-586-1
TSCA TSCA
RTECS Number XS9090000
Melting Point 43° to 46°C
CAS Max % 100.0
Synonym α-Chloro-m-nitrotoluene
CAS Min % 98.5
Health Hazard 1 Danger
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material 3-Nitrobenzyl chloride
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H6ClNO2
Linear Formula O2NC6H4CH2Cl
MDL Number MFCD00007272
CAS 619-23-8
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.