missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzyl bromide, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC206920250

 View more versions of this product

Catalog No. AC206920250



Specifications

Specifications

3-Nitrobenzyl bromide
Glass bottle
55°C to 59°C
3958-57-4
100.0
C7H6BrNO2
MFCD00007271
1-bromomethyl-3-nitrobenzene
C1=CC(=CC(=C1)[N+](=O)[O-])CBr
216.034
216.03
≥98.5%
Authentic
Yellow to Brown
25g
98.5
99%
O2NC6H4CH2Br
05, 334
LNWXALCHPJANMJ-UHFFFAOYSA-N
1-(bromomethyl)-3-nitrobenzene
77568
Crystalline Powder or Crystals

Chemical Identifiers

3958-57-4
216.034
LNWXALCHPJANMJ-UHFFFAOYSA-N
77568
C1=CC(=CC(=C1)[N+](=O)[O-])CBr
C7H6BrNO2
MFCD00007271
1-bromomethyl-3-nitrobenzene
1-(bromomethyl)-3-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents