Please login to your online account to display your discounted pricing

3-Nitrobenzyl bromide, 99%, ACROS Organics™

$41.62 - $149.40

Chemical Identifiers

Chemical Name or Material 3-Nitrobenzyl bromide
CAS 3958-57-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6BrNO2
Linear Formula O2NC6H4CH2Br
Formula Weight 216.03
MDL Number MFCD00007271
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC206920250

  SDS
thermo fisher scientific
AC206920250
Glass bottle 25g Each for $149.40

AC206920050

  SDS
thermo fisher scientific
206920050
Glass bottle 5g Each for $41.62
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Formula Weight 216.03
Assay ≥98.5%
Quantity 25g
Color Yellow to light brown
Melting Point 55° to 59°C
Beilstein 05, 334
CAS Max % 100.0
Chemical Name or Material 3-Nitrobenzyl bromide
Linear Formula O2NC6H4CH2Br
Synonym α-Bromo-m-nitrotoluene
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 98.5
MDL Number MFCD00007271
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Molecular Formula C7H6BrNO2
Assay Percent Range 99%
CAS 3958-57-4
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.