missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzyl Alcohol [Matrix for FABMS and liquid SIMS], TCI America™

Manufacturer:  TCI America S037810G

 View more versions of this product

Catalog No. S037810G



Specifications

Specifications

3-Nitrobenzyl Alcohol [Matrix for FABMS and liquid SIMS]
10g
C7H7NO3
3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
C1=CC(=CC(=C1)[N+](=O)[O-])CO
153.137
153.14
≥99.0% (GC)
Undesignated
619-25-0
MFCD00007273
CWNPOQFCIIFQDM-UHFFFAOYSA-N
(3-nitrophenyl)methanol
69267
Crystal at 20°C

Chemical Identifiers

619-25-0
153.137
CWNPOQFCIIFQDM-UHFFFAOYSA-N
69267
C1=CC(=CC(=C1)[N+](=O)[O-])CO
C7H7NO3
MFCD00007273
3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
(3-nitrophenyl)methanol
Safety and Handling

Safety and Handling

RTECSNumber : DP0657000

TSCA : Yes

Documents