missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzoyl chloride, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 155841000

 View more versions of this product

Catalog No. AC155841000



Specifications

Specifications

3-Nitrobenzoyl chloride
>112°C
Glass bottle
275.0°C to 278.0°C
30.0°C to 35.0°C
98%
97.5
98%
O2NC6H4COCl
09, 381
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl
185.57
185.57
98%
1.4200g/mL
Authentic
1.42
Brown
100g
121-90-4
100.0
C7H4ClNO3
MFCD00007247
3-nitro-benzoyl chloride
NXTNASSYJUXJDV-UHFFFAOYSA-N
3-nitrobenzoyl chloride
8495
Low Melting Solid

Chemical Identifiers

121-90-4
185.57
NXTNASSYJUXJDV-UHFFFAOYSA-N
8495
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl
C7H4ClNO3
MFCD00007247
3-nitro-benzoyl chloride
3-nitrobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-505-9

RTECSNumber : DM6646000

TSCA : TSCA

SDS Documents