Learn More
3-Nitrobenzoyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 155845000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Nitrobenzoyl chloride | |
121-90-4 | |
100.0 | |
275.0°C to 278.0°C | |
Authentic | |
Glass bottle | |
O2NC6H4COCl | |
500g | |
1.42 | |
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol | |
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl | |
185.57 | |
185.57 |
98% | |
97.5 | |
1.4200g/mL | |
>112°C | |
98% | |
C7H4ClNO3 | |
MFCD00007247 | |
09, 381 | |
m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid | |
NXTNASSYJUXJDV-UHFFFAOYSA-N | |
3-nitrobenzoyl chloride | |
8495 | |
98% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-505-9
RTECSNumber : DM6646000
TSCA : TSCA
RUO â Research Use Only