missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Nitrobenzotrifluoride, TCI America™

$51.95 - $342.23

Chemical Identifiers

CAS 98-46-4
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007260
InChI Key WHNAMGUAXHGCHH-UHFFFAOYSA-N
Synonym 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
PubChem CID 7386
IUPAC Name 1-nitro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0280500G View Documents TCI America
N0280500G
500g Each for $342.23
 
N028025G View Documents TCI America
N028025G
25g Each for $51.95
 
Specifications

Chemical Identifiers

98-46-4
191.109
WHNAMGUAXHGCHH-UHFFFAOYSA-N
7386
C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F
C7H4F3NO2
MFCD00007260
3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
1-nitro-3-(trifluoromethyl)benzene

Specifications

203°C
500g
C7H4F3NO2
2306
WHNAMGUAXHGCHH-UHFFFAOYSA-N
1-nitro-3-(trifluoromethyl)benzene
7386
Clear Liquid at 20°C
3-Nitrobenzotrifluoride
Yellow
98-46-4
MFCD00007260
3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F
191.109
191.11
≥96.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0443

RTECSNumber : XT3500000

TSCA : Yes