missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrobenzonitrile, 98%

Chemical Identifiers

CAS 619-24-9
Molecular Formula C7H4N2O2
Formula Weight 148.12
MDL Number MFCD00007194
UN Number UN3439
Beilstein 637674
Synonym 3-Cyanonitrobenzene
Chemical Name or Material 3-Nitrobenzonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1582109 SDS Alfa Aesar™
A1582109
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1582114 SDS Alfa Aesar™
A1582114
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1582130 SDS Alfa Aesar™
A1582130
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

619-24-9
148.12
UN3439
3-Cyanonitrobenzene
C7H4N2O2
MFCD00007194
637674
3-Nitrobenzonitrile

Specifications

Almond-like
114° to 117°C
619-24-9
148.12
UN3439
3-Cyanonitrobenzene
98%
165°C (21mmHg)
10g
C7H4N2O2
MFCD00007194
637674
3-Nitrobenzonitrile

Safety and Handling

GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.

GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : DI4900000

TSCA : TSCA