Learn More
3-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1582130
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitrobenzonitrile | |
114°C to 117°C | |
Almond-like | |
MFCD00007194 | |
UN3439 | |
benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril | |
C1=CC(=CC(=C1)[N+](=O)[O-])C#N | |
148.121 | |
148.12 |
619-24-9 | |
165°C (21 mmHg) | |
C7H4N2O2 | |
250 g | |
637674 | |
RUSAWEHOGCWOPG-UHFFFAOYSA-N | |
3-nitrobenzonitrile | |
12079 | |
98% |
Chemical Identifiers
619-24-9 | |
148.121 | |
RUSAWEHOGCWOPG-UHFFFAOYSA-N | |
12079 | |
C1=CC(=CC(=C1)[N+](=O)[O-])C#N |
C7H4N2O2 | |
MFCD00007194 | |
benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril | |
3-nitrobenzonitrile |
Safety and Handling
GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P310-P302+P352-P312-P330-P363-P501c
H301-H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 210-587-7
RTECSNumber : DI4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only