Please login to your online account to display your discounted pricing

3-Nitrobenzoic Acid, 98%, ACROS Organics™


Quantity 1kg

Manufacturer: thermo fisher scientific  128450010

 View more versions of this product

Catalog No. AC128450010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Powder
Formula Weight 167.12
Quantity 1kg
Color Slightly green to light yellow
Melting Point 139° to 143°C
CAS Max % 100.0
Linear Formula O2NC6H4CO2H
Assay Percent Range 98%
CAS 121-92-6
Molecular Formula C7H5NO4
CAS Min % 97.5
MDL Number MFCD00007251
Infrared Spectrum Authentic
Chemical Name or Material 3-Nitrobenzoic acid
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Nitrobenzoic acid
CAS 121-92-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5NO4
Linear Formula O2NC6H4CO2H
Formula Weight 167.12
MDL Number MFCD00007251
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents