Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Nitrobenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3-Nitrobenzenesulfonyl chloride
CAS 121-51-7
Molecular Formula C6H4ClNO4S
Formula Weight 221.62
MDL Number MFCD00007435
UN Number UN3261
Beilstein 746542
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1767006

 
alfa aesar™
A1767006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1767014

 
alfa aesar™
A1767014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1767022

 
alfa aesar™
A1767022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.62
TSCA TSCA
Assay 98%
Molecular Formula C6H4ClNO4S
MDL Number MFCD00007435
CAS 121-51-7
UN Number UN3261
Beilstein 746542
Flash Point >110°C (230°F)
Quantity 5g
Chemical Name or Material 3-Nitrobenzenesulfonyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 59° to 63°C

Applications
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.