Learn More
3-Nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 415841000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Nitrobenzenesulfonyl chloride | |
121-51-7 | |
100.0 | |
Beige to Yellow | |
96% min. (HPLC) | |
C6H4ClNO4S | |
MFCD00007435 | |
3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u | |
MWWNNNAOGWPTQY-UHFFFAOYSA-N | |
3-nitrobenzenesulfonyl chloride | |
8476 | |
97% |
97% | |
96.0 | |
60.0°C to 65.0°C | |
Authentic | |
Glass bottle | |
SO2ClC6H4NO2 | |
100 g | |
Solubility in water: decomposes | |
C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-] | |
221.62 | |
221.62 | |
Crystalline Powder |
Chemical Identifiers
121-51-7 | |
221.62 | |
MWWNNNAOGWPTQY-UHFFFAOYSA-N | |
8476 | |
C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
C6H4ClNO4S | |
MFCD00007435 | |
3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u | |
3-nitrobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED
GHS Signal Word: Danger
EINECSNumber : 204-476-2
TSCA : TSCA
RUO – Research Use Only